Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137142 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GABRA5 - gamma-aminobutyric acid type A receptor alpha5 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000810.3 | Intron | NP_000801.1 | ||||
NM_001165037.1 | Intron | NP_001158509.1 | ||||
XM_005268258.2 | Intron | XP_005268315.1 | ||||
XM_006720459.2 | Intron | XP_006720522.1 | ||||
XM_017022055.1 | Intron | XP_016877544.1 | ||||
XM_017022056.1 | Intron | XP_016877545.1 |
Set Membership: |
HapMap |