Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605306 | ||||||||||||||||||||||||||||||||
Literature Links: |
CLEC4A PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CLEC4A - C-type lectin domain family 4 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016184.3 | 1651 | Intron | NP_057268.1 | |||
NM_194447.2 | 1651 | Intron | NP_919429.2 | |||
NM_194448.2 | 1651 | Intron | NP_919430.1 | |||
NM_194450.2 | 1651 | Intron | NP_919432.1 | |||
XM_011520684.2 | 1651 | Missense Mutation | AAA,AGA | K,R 11 | XP_011518986.1 | |
XM_017019382.1 | 1651 | Intron | XP_016874871.1 |
POU5F1P3 - POU class 5 homeobox 1 pseudogene 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |