Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTCCTTCCCAGTGGTGGCCATT[A/C]CCCCAGCCTTCTCCATGACGTAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611570 MIM: 603027 | ||||||||||||||||||||
Literature Links: |
FBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBP1 - fructose-bisphosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000507.3 | 1212 | Missense Mutation | NP_000498.2 | |||
NM_001127628.1 | 1212 | Missense Mutation | NP_001121100.1 | |||
XM_006717005.3 | 1212 | Missense Mutation | XP_006717068.1 |
FBP2 - fructose-bisphosphatase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |