Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTCCTCGCTCTCCAGCAGCTTG[A/C]GGTAGGTGGCGATCTCCATATCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608246 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT72 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KRT72 - keratin 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146225.1 | 1071 | Missense Mutation | NP_001139697.1 | |||
NM_001146226.1 | 1071 | Missense Mutation | NP_001139698.1 | |||
NM_080747.2 | 1071 | Missense Mutation | NP_542785.1 | |||
XM_017018833.1 | 1071 | Missense Mutation | XP_016874322.1 | |||
XM_017018834.1 | 1071 | Missense Mutation | XP_016874323.1 | |||
XM_017018835.1 | 1071 | Missense Mutation | XP_016874324.1 | |||
XM_017018836.1 | 1071 | Missense Mutation | XP_016874325.1 | |||
XM_017018837.1 | 1071 | Missense Mutation | XP_016874326.1 |