Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCTCTCCCTAAGATGAAAAGGC[A/G]TCTCTCTGGATTTGGTGGTTCTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606383 MIM: 609124 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR84 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR84 - G protein-coupled receptor 84 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC102724050 - uncharacterized LOC102724050 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF385A - zinc finger protein 385A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130967.2 | Intron | NP_001124439.1 | ||||
NM_001130968.2 | Intron | NP_001124440.1 | ||||
NM_001290001.1 | Intron | NP_001276930.1 | ||||
NM_001290002.1 | Intron | NP_001276931.1 | ||||
NM_001290004.1 | Intron | NP_001276933.1 | ||||
NM_015481.2 | Intron | NP_056296.1 | ||||
XM_005268783.4 | Intron | XP_005268840.1 | ||||
XM_006719342.2 | Intron | XP_006719405.1 | ||||
XM_011538168.2 | Intron | XP_011536470.1 | ||||
XM_011538169.2 | Intron | XP_011536471.1 | ||||
XM_011538170.1 | Intron | XP_011536472.1 | ||||
XM_011538171.2 | Intron | XP_011536473.1 | ||||
XM_017019174.1 | Intron | XP_016874663.1 |