Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610930 MIM: 611052 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ORAI3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ORAI3 - ORAI calcium release-activated calcium modulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD1A - SET domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014712.2 | Intron | NP_055527.1 | ||||
XM_005255723.1 | Intron | XP_005255780.1 | ||||
XM_006721106.3 | Intron | XP_006721169.1 | ||||
XM_017023909.1 | Intron | XP_016879398.1 |
Set Membership: |
HapMap |