Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600396 MIM: 600711 | ||||||||||||||||||||
Literature Links: |
DHX8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHX8 - DEAH-box helicase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302623.1 | 1486 | Intron | NP_001289552.1 | |||
NM_001322216.1 | 1486 | Intron | NP_001309145.1 | |||
NM_001322217.1 | 1486 | Intron | NP_001309146.1 | |||
NM_001322218.1 | 1486 | Intron | NP_001309147.1 | |||
NM_001322219.1 | 1486 | Intron | NP_001309148.1 | |||
NM_001322220.1 | 1486 | Intron | NP_001309149.1 | |||
NM_001322221.1 | 1486 | Intron | NP_001309150.1 | |||
NM_004941.2 | 1486 | Intron | NP_004932.1 |
ETV4 - ETS variant 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079675.2 | 1486 | UTR 3 | NP_001073143.1 | |||
NM_001261437.1 | 1486 | UTR 3 | NP_001248366.1 | |||
NM_001261438.1 | 1486 | UTR 3 | NP_001248367.1 | |||
NM_001261439.1 | 1486 | UTR 3 | NP_001248368.1 | |||
NM_001986.2 | 1486 | UTR 3 | NP_001977.1 | |||
XM_011524514.1 | 1486 | UTR 3 | XP_011522816.1 | |||
XM_017024349.1 | 1486 | UTR 3 | XP_016879838.1 |