Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTTTTTGTTGCCCTCCCCTCCCC[A/T]CTACTGCCCATCTAAGTACTTTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609243 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LRP11 - LDL receptor related protein 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAET1E - retinoic acid early transcript 1E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243325.1 | Intron | NP_001230254.1 | ||||
NM_001243327.1 | Intron | NP_001230256.1 | ||||
NM_001243328.1 | Intron | NP_001230257.1 | ||||
NM_139165.2 | Intron | NP_631904.1 | ||||
XM_011535475.2 | Intron | XP_011533777.1 | ||||
XM_011535476.2 | Intron | XP_011533778.1 | ||||
XM_011535478.2 | Intron | XP_011533780.1 | ||||
XM_011535479.2 | Intron | XP_011533781.1 | ||||
XM_017010285.1 | Intron | XP_016865774.1 | ||||
XM_017010286.1 | Intron | XP_016865775.1 | ||||
XM_017010287.1 | Intron | XP_016865776.1 | ||||
XM_017010288.1 | Intron | XP_016865777.1 |
RAET1E-AS1 - RAET1E antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |