Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600838 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FOXN1 - forkhead box N1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003593.2 | Intron | NP_003584.2 | ||||
XM_005258046.3 | Intron | XP_005258103.1 | ||||
XM_011525358.1 | Intron | XP_011523660.1 | ||||
XM_011525359.1 | Intron | XP_011523661.1 | ||||
XM_011525362.1 | Intron | XP_011523664.1 | ||||
XM_011525367.1 | Intron | XP_011523669.1 | ||||
XM_011525368.2 | Intron | XP_011523670.1 | ||||
XM_011525369.2 | Intron | XP_011523671.1 | ||||
XM_011525370.2 | Intron | XP_011523672.1 | ||||
XM_017025228.1 | Intron | XP_016880717.1 | ||||
XM_017025229.1 | Intron | XP_016880718.1 | ||||
XM_017025230.1 | Intron | XP_016880719.1 | ||||
XM_017025231.1 | Intron | XP_016880720.1 |