Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGTCCATTTGGTCTAAAGTTCAA[C/T]TTAAGTACAATGTTTCTTTGTTAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616980 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf49 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C7orf49 - chromosome 7 open reading frame 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243749.1 | Intron | NP_001230678.1 | ||||
NM_001243751.1 | Intron | NP_001230680.1 | ||||
NM_001243752.1 | Intron | NP_001230681.1 | ||||
NM_001243753.1 | Intron | NP_001230682.1 | ||||
NM_001243754.1 | Intron | NP_001230683.1 | ||||
NM_001243755.1 | Intron | NP_001230684.1 | ||||
NM_001305629.1 | Intron | NP_001292558.1 | ||||
NM_001305630.1 | Intron | NP_001292559.1 | ||||
NM_024033.3 | Intron | NP_076938.2 | ||||
XM_017012589.1 | Intron | XP_016868078.1 | ||||
XM_017012590.1 | Intron | XP_016868079.1 | ||||
XM_017012591.1 | Intron | XP_016868080.1 | ||||
XM_017012592.1 | Intron | XP_016868081.1 | ||||
XM_017012593.1 | Intron | XP_016868082.1 | ||||
XM_017012594.1 | Intron | XP_016868083.1 | ||||
XM_017012595.1 | Intron | XP_016868084.1 |
TMEM140 - transmembrane protein 140 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018295.4 | Intron | NP_060765.4 |
Set Membership: |
HapMap |