Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614282 MIM: 603905 MIM: 600315 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SDF4 - stromal cell derived factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFRSF18 - TNF receptor superfamily member 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFRSF4 - TNF receptor superfamily member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003327.3 | 929 | Silent Mutation | GAA,GAG | E,E 178 | NP_003318.1 | |
XM_011542074.2 | 929 | Silent Mutation | GAA,GAG | E,E 182 | XP_011540376.1 | |
XM_011542075.2 | 929 | Silent Mutation | GAA,GAG | E,E 178 | XP_011540377.1 | |
XM_011542076.2 | 929 | Silent Mutation | GAA,GAG | E,E 182 | XP_011540378.1 | |
XM_011542077.2 | 929 | Silent Mutation | GAA,GAG | E,E 78 | XP_011540379.1 | |
XM_017002231.1 | 929 | Silent Mutation | GAA,GAG | E,E 178 | XP_016857720.1 | |
XM_017002232.1 | 929 | Silent Mutation | GAA,GAG | E,E 178 | XP_016857721.1 |
Set Membership: |
Validated |