Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGCGGGCGCCATAGTGCACATT[A/G]TAGCCGTCGACCAGCACATCTGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616254 | ||||||||||||||||||||
Literature Links: |
CLPB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLPB - ClpB homolog, mitochondrial AAA ATPase chaperonin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258392.2 | 2049 | Silent Mutation | TAC,TAT | Y,Y 583 | NP_001245321.1 | |
NM_001258393.2 | 2049 | Silent Mutation | TAC,TAT | Y,Y 554 | NP_001245322.1 | |
NM_001258394.2 | 2049 | Silent Mutation | TAC,TAT | Y,Y 568 | NP_001245323.1 | |
NM_030813.5 | 2049 | Silent Mutation | TAC,TAT | Y,Y 613 | NP_110440.1 | |
XM_005274320.1 | 2049 | Silent Mutation | TAC,TAT | Y,Y 584 | XP_005274377.1 | |
XM_011545289.2 | 2049 | Intron | XP_011543591.1 |
LOC105369375 - uncharacterized LOC105369375 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |