Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTTTATATGTACGTATACATAT[A/T]CATGGACTAGAGAAGAATCTTCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160994 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPM PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HNRNPM - heterogeneous nuclear ribonucleoprotein M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297418.1 | Intron | NP_001284347.1 | ||||
NM_005968.4 | Intron | NP_005959.2 | ||||
NM_031203.3 | Intron | NP_112480.2 | ||||
XM_005272478.2 | Intron | XP_005272535.1 | ||||
XM_005272479.2 | Intron | XP_005272536.1 | ||||
XM_005272480.2 | Intron | XP_005272537.1 | ||||
XM_005272481.1 | Intron | XP_005272538.1 | ||||
XM_005272483.2 | Intron | XP_005272540.1 | ||||
XM_017026823.1 | Intron | XP_016882312.1 | ||||
XM_017026824.1 | Intron | XP_016882313.1 | ||||
XM_017026825.1 | Intron | XP_016882314.1 | ||||
XM_017026826.1 | Intron | XP_016882315.1 | ||||
XM_017026827.1 | Intron | XP_016882316.1 | ||||
XM_017026828.1 | Intron | XP_016882317.1 | ||||
XM_017026829.1 | Intron | XP_016882318.1 | ||||
XM_017026830.1 | Intron | XP_016882319.1 | ||||
XM_017026831.1 | Intron | XP_016882320.1 | ||||
XM_017026832.1 | Intron | XP_016882321.1 |
Set Membership: |
HapMap |