Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCTTATTTTCAACGGCTTGTAT[C/T]TGTATTTCTCCATATTTTGGAGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604078 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF136 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF136 - zinc finger protein 136 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003437.3 | Intron | NP_003428.1 | ||||
XM_011528269.2 | Intron | XP_011526571.1 | ||||
XM_017027243.1 | Intron | XP_016882732.1 |
ZNF625 - zinc finger protein 625 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF625-ZNF20 - ZNF625-ZNF20 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |