Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCCCAAGCCCAGACACAAACAC[A/G]CATGCTGTCTATACTGCCTGCAAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606237 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGFBRAP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TGFBRAP1 - transforming growth factor beta receptor associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142621.2 | Intron | NP_001136093.1 | ||||
NM_004257.5 | Intron | NP_004248.2 | ||||
XM_011512193.2 | Intron | XP_011510495.1 | ||||
XM_017005339.1 | Intron | XP_016860828.1 |
Set Membership: |
HapMap Validated |