Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604139 MIM: 601773 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAJB2 - DnaJ heat shock protein family (Hsp40) member B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR153-1 - microRNA 153-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPRN - protein tyrosine phosphatase, receptor type N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199763.1 | Intron | NP_001186692.1 | ||||
NM_001199764.1 | Intron | NP_001186693.1 | ||||
NM_002846.3 | Intron | NP_002837.1 | ||||
XM_011511566.1 | Intron | XP_011509868.1 | ||||
XM_017004609.1 | Intron | XP_016860098.1 | ||||
XM_017004610.1 | Intron | XP_016860099.1 | ||||
XM_017004611.1 | Intron | XP_016860100.1 | ||||
XM_017004612.1 | Intron | XP_016860101.1 |
Set Membership: |
HapMap |