Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCCGTTTGTAGCACCACTGGGT[C/G]GGGGGAGACAGGAAGGAGTGAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTS10 - ADAM metallopeptidase with thrombospondin type 1 motif 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282352.1 | 87 | Missense Mutation | CCA,CGA | P,R 18 | NP_001269281.1 | |
NM_030957.3 | 87 | Intron | NP_112219.3 | |||
XM_006722917.3 | 87 | Intron | XP_006722980.1 | |||
XM_017027338.1 | 87 | Intron | XP_016882827.1 | |||
XM_017027339.1 | 87 | Intron | XP_016882828.1 | |||
XM_017027340.1 | 87 | Intron | XP_016882829.1 |