Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATATTTAACCTCTCAAATTGTCT[C/G]TATTGAAGGTTACCTTTTTGACCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614997 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GATAD2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GATAD2A - GATA zinc finger domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300946.1 | Intron | NP_001287875.1 | ||||
NM_017660.3 | Intron | NP_060130.3 | ||||
XM_011528105.1 | Intron | XP_011526407.1 | ||||
XM_017026901.1 | Intron | XP_016882390.1 | ||||
XM_017026902.1 | Intron | XP_016882391.1 | ||||
XM_017026903.1 | Intron | XP_016882392.1 | ||||
XM_017026904.1 | Intron | XP_016882393.1 | ||||
XM_017026905.1 | Intron | XP_016882394.1 | ||||
XM_017026906.1 | Intron | XP_016882395.1 | ||||
XM_017026907.1 | Intron | XP_016882396.1 | ||||
XM_017026908.1 | Intron | XP_016882397.1 | ||||
XM_017026909.1 | Intron | XP_016882398.1 | ||||
XM_017026910.1 | Intron | XP_016882399.1 | ||||
XM_017026911.1 | Intron | XP_016882400.1 |
Set Membership: |
Validated |