Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAATGCTTTAAAGCAAGTTTTCC[C/G]AAGTGGAACCATATATGCCTACTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600543 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERBB4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ERBB4 - erb-b2 receptor tyrosine kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042599.1 | 8735 | UTR 3 | NP_001036064.1 | |||
NM_005235.2 | 8735 | UTR 3 | NP_005226.1 | |||
XM_005246376.2 | 8735 | UTR 3 | XP_005246433.1 | |||
XM_005246377.2 | 8735 | UTR 3 | XP_005246434.1 | |||
XM_006712364.2 | 8735 | UTR 3 | XP_006712427.1 | |||
XM_017003577.1 | 8735 | UTR 3 | XP_016859066.1 | |||
XM_017003578.1 | 8735 | UTR 3 | XP_016859067.1 | |||
XM_017003579.1 | 8735 | UTR 3 | XP_016859068.1 | |||
XM_017003580.1 | 8735 | UTR 3 | XP_016859069.1 | |||
XM_017003581.1 | 8735 | UTR 3 | XP_016859070.1 | |||
XM_017003582.1 | 8735 | UTR 3 | XP_016859071.1 |
Set Membership: |
HapMap |