Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176886 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPRG - protein tyrosine phosphatase, receptor type G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002841.3 | Intron | NP_002832.3 | ||||
XM_005265353.3 | Intron | XP_005265410.1 | ||||
XM_017006961.1 | Intron | XP_016862450.1 | ||||
XM_017006962.1 | Intron | XP_016862451.1 | ||||
XM_017006963.1 | Intron | XP_016862452.1 | ||||
XM_017006964.1 | Intron | XP_016862453.1 |