Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606627 MIM: 603707 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAAM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DAAM2 - dishevelled associated activator of morphogenesis 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOCS1 - molybdenum cofactor synthesis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001075098.3 | Intron | NP_001068566.1 | ||||
NM_005943.5 | Intron | NP_005934.2 | ||||
XM_011514632.2 | Intron | XP_011512934.1 | ||||
XM_011514633.2 | Intron | XP_011512935.1 | ||||
XM_011514634.2 | Intron | XP_011512936.1 | ||||
XM_017010888.1 | Intron | XP_016866377.1 |