Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTCTCCACTTCGCTCTTCTCTG[G/T]CCTTCACCTCTCCTTCATCCATTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608951 | ||||||||||||||||||||
Literature Links: |
CNOT6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNOT6 - CCR4-NOT transcription complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303241.1 | Intron | NP_001290170.1 | ||||
XM_005265953.1 | Intron | XP_005266010.1 | ||||
XM_011534605.2 | Intron | XP_011532907.1 | ||||
XM_011534606.1 | Intron | XP_011532908.1 | ||||
XM_011534607.1 | Intron | XP_011532909.1 | ||||
XM_017009670.1 | Intron | XP_016865159.1 | ||||
XM_017009671.1 | Intron | XP_016865160.1 | ||||
XM_017009672.1 | Intron | XP_016865161.1 |