Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611143 MIM: 606769 MIM: 611983 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM175A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM175A - family with sequence similarity 175 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139076.2 | 2271 | Intron | NP_620775.2 | |||
XM_005263280.4 | 2271 | UTR 3 | XP_005263337.1 | |||
XM_017008694.1 | 2271 | UTR 3 | XP_016864183.1 |
HELQ - helicase, POLQ-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS18C - mitochondrial ribosomal protein S18C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297767.1 | 2271 | UTR 3 | NP_001284696.1 | |||
NM_001297769.1 | 2271 | UTR 3 | NP_001284698.1 | |||
NM_001297770.1 | 2271 | UTR 3 | NP_001284699.1 | |||
NM_016067.3 | 2271 | UTR 3 | NP_057151.1 |
Set Membership: |
HapMap |