Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTCAGTTGTGAGGATCAAGTGA[G/T]TTAATTTACAGTGCTTAGAACACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606022 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUDT9 - nudix hydrolase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001248011.1 | Intron | NP_001234940.1 | ||||
NM_024047.4 | Intron | NP_076952.1 | ||||
NM_198038.2 | Intron | NP_932155.1 | ||||
XM_011532031.2 | Intron | XP_011530333.1 | ||||
XM_011532032.2 | Intron | XP_011530334.1 |
Set Membership: |
HapMap |