Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGTTACAGGGGATTTGTACTTC[C/T]TTTTGAAGATAGCTATTTCATTTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605126 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FHL5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FHL5 - four and a half LIM domains 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170807.2 | Intron | NP_001164278.1 | ||||
NM_001322466.1 | Intron | NP_001309395.1 | ||||
NM_001322467.1 | Intron | NP_001309396.1 | ||||
NM_020482.5 | Intron | NP_065228.4 |
Set Membership: |
HapMap Validated |