Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185620 MIM: 185630 MIM: 185660 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SURF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SURF1 - surfeit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SURF2 - surfeit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278928.1 | 2581 | Intron | NP_001265857.1 | |||
NM_017503.4 | 2581 | Intron | NP_059973.4 |
SURF4 - surfeit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280788.1 | 2581 | UTR 3 | NP_001267717.1 | |||
NM_001280789.1 | 2581 | UTR 3 | NP_001267718.1 | |||
NM_001280790.1 | 2581 | UTR 3 | NP_001267719.1 | |||
NM_001280791.1 | 2581 | UTR 3 | NP_001267720.1 | |||
NM_001280792.1 | 2581 | UTR 3 | NP_001267721.1 | |||
NM_001280793.1 | 2581 | UTR 3 | NP_001267722.1 | |||
NM_033161.3 | 2581 | UTR 3 | NP_149351.1 | |||
XM_011518943.1 | 2581 | UTR 3 | XP_011517245.1 | |||
XM_011518944.2 | 2581 | UTR 3 | XP_011517246.1 | |||
XM_011518945.2 | 2581 | UTR 3 | XP_011517247.1 | |||
XM_017015062.1 | 2581 | UTR 3 | XP_016870551.1 |
Set Membership: |
HapMap |