Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTCAAAGAAAGAAAAAGAAGTA[C/T]AGAAAAGAGAGAAAAACGTCAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602325 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4G2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF4G2 - eukaryotic translation initiation factor 4 gamma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042559.2 | Intron | NP_001036024.3 | ||||
NM_001172705.1 | Intron | NP_001166176.1 | ||||
NM_001418.3 | Intron | NP_001409.3 |
LOC101928053 - uncharacterized LOC101928053 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105376545 - uncharacterized LOC105376545 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD97 - small nucleolar RNA, C/D box 97 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |