Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGTCTGCTGAAGGCCCTGCCCC[A/G]CTGCCTGCTGGATGTCAGCCCCTTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606491 MIM: 603048 MIM: 614243 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOSC4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
EXOSC4 - exosome component 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019037.2 | Intron | NP_061910.1 | ||||
XM_011517134.2 | Intron | XP_011515436.1 |
GPAA1 - glycosylphosphatidylinositol anchor attachment 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6847 - microRNA 6847 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OPLAH - 5-oxoprolinase (ATP-hydrolysing) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |