Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACAGCTTAGGAAAGAATGAAGAT[A/G]GGTTTAATCCACTGAGTCCAGATCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610770 MIM: 615921 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL17 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLHL17 - kelch like family member 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOC2L - NOC2 like nucleolar associated transcriptional repressor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PERM1 - PPARGC1 and ESRR induced regulator, muscle 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLEKHN1 - pleckstrin homology domain containing N1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160184.1 | Intron | NP_001153656.1 | ||||
NM_032129.2 | Intron | NP_115505.2 | ||||
XM_006710944.3 | Intron | XP_006711007.2 | ||||
XM_011542248.2 | Intron | XP_011540550.2 | ||||
XM_017002474.1 | Intron | XP_016857963.1 | ||||
XM_017002475.1 | Intron | XP_016857964.1 | ||||
XM_017002476.1 | Intron | XP_016857965.1 | ||||
XM_017002477.1 | Intron | XP_016857966.1 | ||||
XM_017002478.1 | Intron | XP_016857967.1 | ||||
XM_017002479.1 | Intron | XP_016857968.1 |