Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300522 MIM: 314690 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQSEC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IQSEC2 - IQ motif and Sec7 domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111125.2 | 4436 | Intron | NP_001104595.1 | |||
NM_001243197.1 | 4436 | Intron | NP_001230126.1 | |||
NM_015075.1 | 4436 | Intron | NP_055890.1 | |||
XM_006724579.2 | 4436 | Intron | XP_006724642.1 | |||
XM_006724580.3 | 4436 | Intron | XP_006724643.1 | |||
XM_006724581.3 | 4436 | Intron | XP_006724644.1 | |||
XM_006724582.3 | 4436 | UTR 3 | XP_006724645.1 | |||
XM_006724583.3 | 4436 | Intron | XP_006724646.1 | |||
XM_006724584.2 | 4436 | Intron | XP_006724647.1 | |||
XM_011530773.2 | 4436 | Intron | XP_011529075.1 | |||
XM_011530774.2 | 4436 | Intron | XP_011529076.1 | |||
XM_011530776.1 | 4436 | Intron | XP_011529078.1 | |||
XM_011530777.1 | 4436 | Intron | XP_011529079.1 | |||
XM_017029359.1 | 4436 | Intron | XP_016884848.1 | |||
XM_017029360.1 | 4436 | Intron | XP_016884849.1 |
KDM5C - lysine demethylase 5C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |