Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMEL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MMEL1 - membrane metallo-endopeptidase-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033467.3 | 501 | Silent Mutation | CCA,CCC | P,P 7 | NP_258428.2 | |
XM_011542122.2 | 501 | Intron | XP_011540424.1 | |||
XM_017002310.1 | 501 | Silent Mutation | CCA,CCC | P,P 7 | XP_016857799.1 | |
XM_017002311.1 | 501 | Silent Mutation | CCA,CCC | P,P 7 | XP_016857800.1 | |
XM_017002312.1 | 501 | Silent Mutation | CCA,CCC | P,P 7 | XP_016857801.1 | |
XM_017002313.1 | 501 | Silent Mutation | CCA,CCC | P,P 7 | XP_016857802.1 | |
XM_017002314.1 | 501 | Silent Mutation | CCA,CCC | P,P 7 | XP_016857803.1 | |
XM_017002315.1 | 501 | Intron | XP_016857804.1 |
TTC34 - tetratricopeptide repeat domain 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |