Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610465 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ACSF2 - acyl-CoA synthetase family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288968.1 | Intron | NP_001275897.1 | ||||
NM_001288969.1 | Intron | NP_001275898.1 | ||||
NM_001288970.1 | Intron | NP_001275899.1 | ||||
NM_001288971.1 | Intron | NP_001275900.1 | ||||
NM_001288972.1 | Intron | NP_001275901.1 | ||||
NM_025149.5 | Intron | NP_079425.3 | ||||
XM_006722110.3 | Intron | XP_006722173.1 | ||||
XM_011525294.1 | Intron | XP_011523596.1 |
Set Membership: |
HapMap Validated |