Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608025 | ||||||||||||||||||||
Literature Links: |
NBAS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NBAS - neuroblastoma amplified sequence | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015909.3 | Intron | NP_056993.2 | ||||
XM_011510357.2 | Intron | XP_011508659.1 | ||||
XM_011510358.2 | Intron | XP_011508660.1 | ||||
XM_011510359.2 | Intron | XP_011508661.1 | ||||
XM_011510360.2 | Intron | XP_011508662.1 | ||||
XM_011510361.2 | Intron | XP_011508663.1 | ||||
XM_017004317.1 | Intron | XP_016859806.1 |