Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCAATCACAGAAAAGCGTGCTT[A/G]TAGTTTAAAACATGTATTACAATGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603102 MIM: 604026 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPD PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CPD - carboxypeptidase D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GOSR1 - golgi SNAP receptor complex member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007024.1 | Intron | NP_001007025.1 | ||||
NM_001007025.1 | Intron | NP_001007026.1 | ||||
NM_004871.2 | Intron | NP_004862.1 | ||||
XM_005258070.4 | Intron | XP_005258127.1 | ||||
XM_005258071.4 | Intron | XP_005258128.1 | ||||
XM_005258072.2 | Intron | XP_005258129.1 | ||||
XM_017025374.1 | Intron | XP_016880863.1 | ||||
XM_017025375.1 | Intron | XP_016880864.1 | ||||
XM_017025376.1 | Intron | XP_016880865.1 | ||||
XM_017025377.1 | Intron | XP_016880866.1 |
Set Membership: |
HapMap |