Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAATGGAAACGGAAGATTTTGG[A/G]CGGTTATTCCTACCCAAGTCCTTAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601511 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STAT5A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
STAT5A - signal transducer and activator of transcription 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288718.1 | Intron | NP_001275647.1 | ||||
NM_001288719.1 | Intron | NP_001275648.1 | ||||
NM_001288720.1 | Intron | NP_001275649.1 | ||||
NM_003152.3 | Intron | NP_003143.2 | ||||
XM_005257624.3 | Intron | XP_005257681.1 |
Set Membership: |
HapMap |