Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607570 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX40 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DHX40 - DEAH-box helicase 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166301.1 | Intron | NP_001159773.1 | ||||
NM_024612.4 | Intron | NP_078888.4 | ||||
XM_006722083.2 | Intron | XP_006722146.1 | ||||
XM_011525253.2 | Intron | XP_011523555.1 | ||||
XM_011525254.2 | Intron | XP_011523556.1 | ||||
XM_017025080.1 | Intron | XP_016880569.1 | ||||
XM_017025081.1 | Intron | XP_016880570.1 | ||||
XM_017025082.1 | Intron | XP_016880571.1 | ||||
XM_017025083.1 | Intron | XP_016880572.1 |
Set Membership: |
HapMap |