Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615769 MIM: 612497 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM169A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FAM169A - family with sequence similarity 169 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015566.2 | Intron | NP_056381.1 | ||||
XM_005248480.2 | Intron | XP_005248537.1 | ||||
XM_011543306.2 | Intron | XP_011541608.1 | ||||
XM_011543307.2 | Intron | XP_011541609.1 | ||||
XM_017009332.1 | Intron | XP_016864821.1 | ||||
XM_017009333.1 | Intron | XP_016864822.1 |
NSA2 - NSA2, ribosome biogenesis homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |