Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 605575 MIM: 616148 | |||||||||||||||||||||||||||||
Literature Links: |
SMC4 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
SMC4 - structural maintenance of chromosomes 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002800.2 | 3862 | Intron | NP_001002800.1 | |||
NM_001288753.1 | 3862 | Intron | NP_001275682.1 | |||
NM_005496.3 | 3862 | Intron | NP_005487.3 | |||
XM_006713459.3 | 3862 | Intron | XP_006713522.1 | |||
XM_011512311.2 | 3862 | Intron | XP_011510613.1 | |||
XM_011512312.2 | 3862 | Intron | XP_011510614.1 | |||
XM_017005491.1 | 3862 | Intron | XP_016860980.1 |
TRIM59 - tripartite motif containing 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173084.2 | 3862 | UTR 3 | NP_775107.1 |