Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610005 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNIK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNIK - TRAF2 and NCK interacting kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161560.2 | 4452 | UTR 3 | NP_001155032.1 | |||
NM_001161561.2 | 4452 | UTR 3 | NP_001155033.1 | |||
NM_001161562.2 | 4452 | UTR 3 | NP_001155034.1 | |||
NM_001161563.2 | 4452 | UTR 3 | NP_001155035.1 | |||
NM_001161564.2 | 4452 | UTR 3 | NP_001155036.1 | |||
NM_001161565.2 | 4452 | UTR 3 | NP_001155037.1 | |||
NM_001161566.2 | 4452 | UTR 3 | NP_001155038.1 | |||
NM_015028.3 | 4452 | UTR 3 | NP_055843.1 |