Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138160 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC2A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC2A2 - solute carrier family 2 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000340.1 | 1741 | Silent Mutation | CTG,GTG | L,V 478 | NP_000331.1 | |
NM_001278658.1 | 1741 | Silent Mutation | CTG,GTG | L,V 359 | NP_001265587.1 | |
NM_001278659.1 | 1741 | Silent Mutation | CTG,GTG | L,V 305 | NP_001265588.1 | |
XM_011513087.2 | 1741 | Missense Mutation | CTG,GTG | L,V 463 | XP_011511389.1 | |
XM_011513089.2 | 1741 | Silent Mutation | CTG,GTG | L,V 305 | XP_011511391.1 |