Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300296 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00629 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LINC00629 - long intergenic non-protein coding RNA 629 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLAC1 - placenta specific 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316887.1 | 1039 | UTR 3 | NP_001303816.1 | |||
NM_001316888.1 | 1039 | UTR 3 | NP_001303817.1 | |||
NM_001316889.1 | 1039 | UTR 3 | NP_001303818.1 | |||
NM_021796.3 | 1039 | UTR 3 | NP_068568.1 | |||
XM_011531257.2 | 1039 | UTR 3 | XP_011529559.1 | |||
XM_011531260.2 | 1039 | UTR 3 | XP_011529562.1 | |||
XM_017029236.1 | 1039 | UTR 3 | XP_016884725.1 |