Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAAACACCCCTGCCTGCTCTCCG[C/T]TTCCTTCTCCACCCCAGCTTGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300381 | ||||||||||||||||||||
Literature Links: |
ZNF185 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF185 - zinc finger protein 185 (LIM domain) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178106.1 | Intron | NP_001171577.1 | ||||
NM_001178107.1 | Intron | NP_001171578.1 | ||||
NM_001178108.1 | Intron | NP_001171579.1 | ||||
NM_001178109.1 | Intron | NP_001171580.1 | ||||
NM_001178110.1 | Intron | NP_001171581.1 | ||||
NM_001178113.1 | Intron | NP_001171584.1 | ||||
NM_001178115.1 | Intron | NP_001171586.1 | ||||
NM_007150.3 | Intron | NP_009081.2 |