Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCATGTCTAAATCATTTTTCTA[A/G]TTATGTCCTATTAAGAAACTACTAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602173 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100128164 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100128164 - four and a half LIM domains 1 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEC62 - SEC62 homolog, preprotein translocation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003262.3 | Intron | NP_003253.1 | ||||
XM_011513114.2 | Intron | XP_011511416.1 |