Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603184 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDK8 - cyclin dependent kinase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260.2 | Intron | NP_001251.1 | ||||
NM_001318368.1 | Intron | NP_001305297.1 | ||||
XM_005266216.4 | Intron | XP_005266273.1 | ||||
XM_011534864.1 | Intron | XP_011533166.1 | ||||
XM_011534865.2 | Intron | XP_011533167.1 | ||||
XM_017020317.1 | Intron | XP_016875806.1 | ||||
XM_017020318.1 | Intron | XP_016875807.1 |
Set Membership: |
HapMap |