Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTGTGTACCACCATGCCCAGCT[G/T]ATTGATTGATTGATTGATTGAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609394 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPC24 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPC24 - SPC24, NDC80 kinetochore complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317031.1 | Intron | NP_001303960.1 | ||||
NM_001317032.1 | Intron | NP_001303961.1 | ||||
NM_001317033.1 | Intron | NP_001303962.1 | ||||
NM_182513.3 | Intron | NP_872319.1 | ||||
XM_005259753.3 | Intron | XP_005259810.1 | ||||
XM_011527702.1 | Intron | XP_011526004.1 |