Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118502 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRNA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHRNA2 - cholinergic receptor nicotinic alpha 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000742.3 | Intron | NP_000733.2 | ||||
NM_001282455.1 | Intron | NP_001269384.1 | ||||
XM_005273397.2 | Intron | XP_005273454.1 | ||||
XM_006716282.1 | Intron | XP_006716345.1 | ||||
XM_011544388.1 | Intron | XP_011542690.1 | ||||
XM_011544389.1 | Intron | XP_011542691.1 | ||||
XM_017013003.1 | Intron | XP_016868492.1 | ||||
XM_017013004.1 | Intron | XP_016868493.1 |
Set Membership: |
HapMap |