Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCCTTCCGCTCACCGCCGCCGCC[C/T]CGGCCACACCCCCTAGAGGGGCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105379298 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105379298 - uncharacterized LOC105379298 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZDHHC2 - zinc finger DHHC-type containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016353.4 | Intron | NP_057437.1 | ||||
XM_011544544.2 | Intron | XP_011542846.1 | ||||
XM_011544545.2 | Intron | XP_011542847.1 | ||||
XM_011544546.2 | Intron | XP_011542848.1 | ||||
XM_011544547.2 | Intron | XP_011542849.1 | ||||
XM_011544548.2 | Intron | XP_011542850.1 | ||||
XM_011544549.2 | Intron | XP_011542851.1 |