Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607588 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPIL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPIL2 - peptidylprolyl isomerase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317996.1 | Intron | NP_001304925.1 | ||||
NM_014337.3 | Intron | NP_055152.1 | ||||
NM_148175.2 | Intron | NP_680480.1 | ||||
NM_148176.2 | Intron | NP_680481.1 | ||||
XM_005261448.3 | Intron | XP_005261505.1 | ||||
XM_011530041.2 | Intron | XP_011528343.1 | ||||
XM_011530042.2 | Intron | XP_011528344.1 | ||||
XM_011530043.2 | Intron | XP_011528345.1 | ||||
XM_011530044.2 | Intron | XP_011528346.1 | ||||
XM_011530046.2 | Intron | XP_011528348.1 | ||||
XM_011530047.2 | Intron | XP_011528349.1 | ||||
XM_011530049.2 | Intron | XP_011528351.1 | ||||
XM_011530050.2 | Intron | XP_011528352.1 | ||||
XM_011530051.2 | Intron | XP_011528353.1 | ||||
XM_017028705.1 | Intron | XP_016884194.1 | ||||
XM_017028706.1 | Intron | XP_016884195.1 |
Set Membership: |
HapMap |