Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614235 MIM: 193001 | ||||||||||||||||||||
Literature Links: |
PDZD8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDZD8 - PDZ domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173791.4 | 6372 | UTR 3 | NP_776152.1 | |||
XM_005269518.3 | 6372 | Intron | XP_005269575.1 | |||
XM_011539265.1 | 6372 | Intron | XP_011537567.1 | |||
XM_011539266.2 | 6372 | Intron | XP_011537568.1 |
SLC18A2 - solute carrier family 18 member A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |