Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTGAAAGTTAGGGACTTTGACT[A/G]TGTGTGGAACCACTTGAATTCATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615164 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKIRIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AKIRIN1 - akirin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136275.1 | Intron | NP_001129747.1 | ||||
NM_024595.2 | Intron | NP_078871.1 |
Set Membership: |
Validated |